Assay Details
Gene Symbol: Ndst4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022565.2 | NP_072087.2 | 5 - 6 | 2062 | 103 | |
GenBank mRNA | AB036838.1 | - | 4 - 5 | 1512 | 103 | |
AK140992.1 | - | 5 - 6 | 2063 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
