Assay Details
Gene Symbol: Ndst4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022565.2 | NP_072087.2 | 6 - 7 | 2192 | 119 | |
GenBank mRNA | AK140992.1 | - | 6 - 7 | 2193 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
