| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1400006783.mm.1 |
NM_026401.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400006783.mm.1 |
NM_026401.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400006783.mm.1 |
TC1400000827.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400006782.mm.1 |
NM_026401.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400006782.mm.1 |
NM_026401.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400006782.mm.1 |
TC1400000827.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400006786.mm.1 |
NM_026401.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400006786.mm.1 |
NM_026401.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400006786.mm.1 |
TC1400000827.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400006785.mm.1 |
NM_026401.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400006785.mm.1 |
NM_026401.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400006785.mm.1 |
TC1400000827.mm.1 |
MTA TC |