Assay Details
| Gene Symbol: Acbd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133225.3 | NP_573488.2 | 2 - 3 | 476 | 65 | |
| GenBank mRNA | AF022770.3 | - | 2 - 3 | 477 | 65 | |
| AK030371.1 | - | 2 - 3 | 477 | 65 | ||
| AK167272.1 | - | 2 - 3 | 458 | 65 | ||
| AK168799.1 | - | 2 - 3 | 448 | 65 | ||
| AK169505.1 | - | 1 - 2 | 3450 | 65 | ||
| BC050121.1 | - | 2 - 3 | 447 | 65 | ||
| BC060602.1 | - | 2 - 3 | 431 | 5706182 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acbd3
Genomic Map