Assay Details
| Gene Symbol: Tmem63a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144794.2 | NP_659043.1 | 7 - 8 | 616 | 79 | |
| GenBank mRNA | AK049360.1 | - | 7 - 8 | 610 | 79 | |
| AK144899.1 | - | 7 - 8 | 669 | 79 | ||
| AK170559.1 | - | 7 - 8 | 632 | 79 | ||
| AK173027.1 | - | 7 - 8 | 609 | 79 | ||
| AK215121.1 | - | 2 - 3 | 191 | 79 | ||
| BC014795.1 | - | 7 - 8 | 589 | 3961992 | 79 | |
| BC019442.1 | - | 7 - 8 | 616 | 4036849 | 79 | |
More Information
Set Membership: |
|

Tmem63a
Genomic Map