Assay Details
| Gene Symbol: Gmpr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177992.2 | NP_818773.1 | 1 - 2 | 148 | 97 | |
| GenBank mRNA | AK017821.1 | - | 1 - 2 | 443 | 97 | |
| AK031004.1 | - | 1 - 2 | 150 | 97 | ||
| AK075772.1 | - | 1 - 2 | 106 | 97 | ||
| BC003886.1 | - | 1 - 2 | 107 | 3590411 | 97 | |
| BC024109.1 | - | 1 - 2 | 104 | 5149353 | 97 | |
| BY099107.1 | - | 1 - 2 | 149 | 97 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK031004.1, AK075772.1, BC003886.1, BC024109.1, NM_177992.2 transcripts.
Set Membership: |
|

Gmpr2
Genomic Map