Assay Details
| Gene Symbol: Gmpr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177992.2 | NP_818773.1 | 4 - 5 | 451 | 80 | |
| GenBank mRNA | AK031004.1 | - | 4 - 5 | 453 | 80 | |
| AK075772.1 | - | 4 - 5 | 409 | 80 | ||
| BC003886.1 | - | 4 - 5 | 410 | 3590411 | 80 | |
| BC024109.1 | - | 4 - 5 | 407 | 5149353 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gmpr2
Genomic Map