Assay Details
| Gene Symbol: Mttp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163457.1 | NP_001156929.1 | 11 - 12 | 1620 | 73 | |
| NM_008642.2 | NP_032668.2 | 9 - 10 | 1310 | 73 | ||
| GenBank mRNA | AK136278.1 | - | 11 - 12 | 1620 | 73 | |
| AK146553.1 | - | 9 - 10 | 1311 | 73 | ||
| BC012686.1 | - | 9 - 10 | 1270 | 4163901 | 73 | |
| EU553486.1 | - | 9 - 10 | 1278 | 73 | ||
| L47970.1 | - | 9 - 10 | 1257 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mttp
Genomic Map