Assay Details
| Gene Symbol: Pvr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027514.2 | NP_081790.1 | 2 - 3 | 535 | 63 | |
| GenBank mRNA | AB195430.1 | - | 2 - 3 | 492 | 63 | |
| AJ563420.1 | - | 2 - 3 | 423 | 63 | ||
| AK014446.1 | - | 2 - 3 | 506 | 63 | ||
| AK038747.1 | - | 2 - 3 | 506 | 63 | ||
| AK078344.1 | - | 2 - 3 | 502 | 63 | ||
| AK080753.1 | - | 2 - 3 | 506 | 63 | ||
| AK165873.1 | - | 2 - 3 | 525 | 63 | ||
| BC013673.1 | - | 2 - 3 | 492 | 4188634 | 63 | |
| BC032283.1 | - | 2 - 3 | 495 | 5363187 | 63 | |
| BC138219.1 | - | 2 - 3 | 480 | 8861238 | 63 | |
| BC138220.1 | - | 2 - 3 | 480 | 8861239 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pvr
Genomic Map