Assay Details
| Gene Symbol: Apoc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277944.1 | NP_001264873.1 | 2 - 3 | 241 | 62 | |
| GenBank mRNA | AK146468.1 | - | 1 - 2 | 42 | 62 | |
| BC024697.1 | - | 2 - 3 | 241 | 4195301 | 62 | |
| BG085960.2 | - | 1 - 2 | 84 | 62 | ||
| CB949564.1 | - | 1 - 2 | 147 | 62 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK146468.1, BC024697.1, NM_001277944.1 transcripts.
Set Membership: |
|

Apoc2
Genomic Map