Assay Details
| Gene Symbol: Ckm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007710.2 | NP_031736.1 | 4 - 5 | 550 | 72 | |
| GenBank mRNA | AK009950.1 | - | 4 - 5 | 551 | 72 | |
| AK137460.1 | - | 552 | 72 | |||
| BC132424.1 | - | 4 - 5 | 548 | 40130701 | 72 | |
| BC132426.1 | - | 4 - 5 | 548 | 40130703 | 72 | |
| X03233.1 | - | 4 - 5 | 550 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24993221 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24993220 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24993219 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P380807 | BC132426.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P380807 | X03233.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P380807 | BC132424.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P380807 | AK009950.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P380807 | AK137460.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P380807 | BC132426.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P380807 | X03233.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P380807 | BC132424.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P380807 | AK009950.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P380807 | AK137460.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 953100 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 496534 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 803718 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 520547 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 875303 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 61560 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 204140 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 53449 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 18445 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P154316 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P123335 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P380807 | BC132426.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P380807 | X03233.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P380807 | BC132424.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P380807 | AK009950.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P380807 | AK137460.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P380807 | BC132426.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P380807 | X03233.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P380807 | BC132424.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P380807 | AK009950.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P380807 | AK137460.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P380807 | BC132426.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P380807 | X03233.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P380807 | BC132424.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P380807 | AK009950.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P380807 | AK137460.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P380807 | BC132426.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P380807 | X03233.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P380807 | BC132424.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P380807 | AK009950.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P380807 | AK137460.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24993159 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24993177 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24993163 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24993173 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24993174 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24993170 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24993176 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24993160 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24993165 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24993158 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24993168 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24993161 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24993179 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P123335 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P154316 | - | - |
| Whole Mouse Genome G4122A | A_51_P380807 | BC132426.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P380807 | X03233.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P380807 | BC132424.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P380807 | AK009950.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P380807 | AK137460.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P380807 | BC132426.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P380807 | X03233.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P380807 | BC132424.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P380807 | AK009950.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P380807 | AK137460.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P113479 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128187 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P123335 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P154316 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06276000 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06145227 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06145215 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06276006 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24993219 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24993220 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24993221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24993214 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06368797 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24993143 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06055474 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24993222 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24993200 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05920371 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06145215 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06629803 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06145213 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06145223 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06603391 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06055475 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24993211 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24993205 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06276004 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06276002 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24993165 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24993146 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24993168 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06356734 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24993174 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24993132 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06055478 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24993156 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06250702 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06250700 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06473689 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24993190 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24993202 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24993170 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24993226 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24993224 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06527229 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24993194 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05890592 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06238989 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05983484 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24993216 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24993221 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24993215 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06629801 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24993220 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24993214 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24993153 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24993176 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06629802 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05953178 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24993173 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24993130 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24993228 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06629791 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24993159 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06020755 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24993213 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05890589 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24993179 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24993138 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06244918 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10550703 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10550699 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10550702 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10560507 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10560509 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10550701 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10550700 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10550705 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10560506 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10550704 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417614_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10560505 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10550698 | - | - |
| Mouse Genome 430 2.0 Array | 1444643_at | - | - |
| Mouse Genome 430 2.0 Array | 1417614_at | - | - |
| Mouse Genome 430 2.0 Array | 1444643_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1430937_at | - | - |
| Mouse Genome 430 2.0 Array | 1417614_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1430937_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12715_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12715_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700002395.mm.1 | TC0700000301.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700002395.mm.1 | NM_007710.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700002395.mm.1 | NM_007710.2 | Refseq |
| Mouse Expression Set 430 | 1417614_PM_at | - | - |
| Mouse Expression Set 430 | 1417614_at | - | - |
| Mouse Expression Set 430 | 1444643_at | - | - |
| Mouse Expression Set 430 | 1444643_PM_at | - | - |
| Murine Genome U74v2 Set | 160175_at | - | - |
| Murine Genome U74v2 Set | 160175_PM_at | - | - |
| Murine Genome U74v2 Set | 161569_PM_f_at | - | - |
| Murine Genome U74v2 Set | 161569_f_at | - | - |
| Murine Genome U74v2 Set | 170247_PM_r_at | - | - |
| Murine Genome U74v2 Set | 170247_r_at | - | - |
| Murine Genome U74v2 Set | 171149_r_at | - | - |
| Murine Genome U74v2 Set | 171149_PM_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417614_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417614_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417614_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417614_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1450524 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001450524 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2608804 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2608804 | - | - |
| mouse-6 v1.1 (Illumina) | scl32994.8.1_63-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1450524 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl32994.8.1_63-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1450524 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2608804 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2608804 | - | - |

Ckm
Genomic Map