Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700002091.mm.1 |
TC0700000283.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700002091.mm.1 |
NM_001159671.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002091.mm.1 |
NM_001159671.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002091.mm.1 |
NM_031255.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002091.mm.1 |
NM_031255.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002094.mm.1 |
TC0700000283.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700002094.mm.1 |
NM_031255.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002094.mm.1 |
NM_031255.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002095.mm.1 |
TC0700000283.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700002095.mm.1 |
NM_031255.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002095.mm.1 |
NM_031255.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002093.mm.1 |
TC0700000283.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700002093.mm.1 |
NM_001159671.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002093.mm.1 |
NM_001159671.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002093.mm.1 |
NM_031255.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002093.mm.1 |
NM_031255.2 |
Refseq |