Assay Details
| Gene Symbol: Slc44a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145394.3 | NP_663369.2 | 11 - 12 | 1550 | 95 | |
| GenBank mRNA | BC010552.1 | - | 10 - 11 | 1318 | 3582543 | 95 |
| BC025548.1 | - | 11 - 12 | 1548 | 5320841 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc44a3
Genomic Map