Assay Details
| Gene Symbol: Aifm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290364.1 | NP_001277293.1 | 11 - 12 | 1370 | 72 | |
| NM_012019.3 | NP_036149.1 | 11 - 12 | 1382 | 72 | ||
| GenBank mRNA | AF100927.1 | - | 11 - 12 | 1208 | 72 | |
| AK133764.1 | - | 11 - 12 | 1222 | 72 | ||
| AK165762.1 | - | 1 - 2 | 173 | 72 | ||
| BC003292.1 | - | 11 - 12 | 1245 | 3500037 | 72 | |
| BC024882.1 | - | 11 - 12 | 1866 | 72 | ||
| DQ016499.1 | - | 10 - 11 | 2509 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Aifm1
Genomic Map