Assay Details
| Gene Symbol: Aifm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290364.1 | NP_001277293.1 | 8 - 9 | 1068 | 79 | |
| NM_012019.3 | NP_036149.1 | 8 - 9 | 1080 | 79 | ||
| GenBank mRNA | AF100927.1 | - | 8 - 9 | 906 | 79 | |
| AK133764.1 | - | 8 - 9 | 920 | 79 | ||
| BC003292.1 | - | 8 - 9 | 943 | 3500037 | 79 | |
| BC024882.1 | - | 8 - 9 | 1564 | 79 | ||
| DQ016499.1 | - | 8 - 9 | 943 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Aifm1
Genomic Map