Assay Details
| Gene Symbol: D1Pas1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033077.3 | NP_149068.1 | 1 | 633 | 61 | |
| GenBank mRNA | AK029542.1 | - | 1 | 633 | 61 | |
| BC139286.1 | - | 1 | 628 | 8862308 | 61 | |
| BC139287.1 | - | 1 | 628 | 8862309 | 61 | |
| J04847.1 | - | 1 | 173 | 61 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

D1Pas1
Genomic Map