Assay Details
| Gene Symbol: Rbm48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172991.4 | NP_766579.3 | 3 - 4 | 926 | 123 | |
| GenBank mRNA | AK047811.1 | - | 3 - 4 | 458 | 123 | |
| BC029549.1 | - | 3 - 4 | 790 | 5012510 | 123 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rbm48
Genomic Map