Assay Details
| Gene Symbol: Kif26b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161665.1 | NP_001155137.1 | 5 - 6 | 1789 | 71 | |
| GenBank mRNA | AB355846.1 | - | 5 - 6 | 1789 | 71 | |
| BC068141.1 | - | 2 - 3 | 601 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kif26b
Genomic Map