Assay Details
Gene Symbol: 1700003H04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_015460.1 | 3 - 4 | 449 | 104 | ||
GenBank mRNA | AK015989.1 | - | 3 - 4 | 450 | 104 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 1700006A11 gene | 1700006A11Rik | -139.9kb | Sense | Chr. 3 - 124400984 - 124426028 on NCBI Build GRCm38 | |
Downstream | N-deacetylase/N-sulfotransferase (heparin glucosaminyl) 4 | Ndst4 | 4930439H17Rik | 825.5kb | Anti-sense | Chr. 3 - 125404076 - 125728351 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2141826 | AK016589.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 607410 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P129568 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P114003 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P129568 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P114003 | - | - |
Whole Mouse Genome G4122A | A_51_P430754 | AK016589.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P606411 | AK016589.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P606411 | BC100480.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P606411 | NR_015460.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P606411 | AK005639.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P606411 | AK015989.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P129568 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P114003 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493695 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02341147 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02822306 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493494 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493498 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493690 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493691 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493489 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493700 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02418446 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02915079 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493621 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493655 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02769719 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493667 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02410297 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493633 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493666 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493620 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493679 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493639 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493612 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493674 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02410291 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493662 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493645 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493500 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493714 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493702 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493717 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493626 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02947846 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493683 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493614 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493699 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493652 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493492 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493658 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493720 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493664 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493490 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493641 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493687 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02467585 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493726 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493636 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493681 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493628 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493723 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493647 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493643 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493660 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493611 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493649 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493629 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493686 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493671 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493627 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493625 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02490092 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493718 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493661 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493496 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493657 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493672 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493637 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493617 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493665 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493716 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493669 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493631 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493697 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22493678 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10501956 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10501958 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10501955 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10501957 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1432354_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1429482_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10501953 | - | - |
Mouse Genome 430 2.0 Array | 1429482_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1429482_at | - | - |
Mouse Genome 430 2.0 Array | 1432354_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1432354_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0300022850.mm.1 | NR_015460.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300022850.mm.1 | NR_015460.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300022850.mm.1 | TC0300002888.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0300022843.mm.1 | NR_015460.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300022843.mm.1 | NR_015460.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300022843.mm.1 | TC0300002888.mm.1 | MTA TC |
Mouse Expression Set 430 | 1432354_PM_at | - | - |
Mouse Expression Set 430 | 1429482_PM_at | - | - |
Mouse Expression Set 430 | 1429482_at | - | - |
Mouse Expression Set 430 | 1432354_at | - | - |
Murine Genome U74v2 Set | 165896_PM_i_at | - | - |
Murine Genome U74v2 Set | 165896_i_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1429482_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1432354_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1429482_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1432354_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 728355 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1217966 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1217966 | - | - |
