Assay Details
| Gene Symbol: 1700061I17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038029.1 | 7 - 8 | 1371 | 100 | ||
| GenBank mRNA | AK006854.1 | - | 7 - 8 | 1373 | 100 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 4930455H04 gene | 4930455H04Rik | -76.7kb | Anti-sense | Chr. 3 - 116968267 - 116984406 on NCBI Build GRCm38 | |
| Downstream | lipid phosphate phosphatase-related protein type 4 | Lppr4 |
A330086D10, D3Bwg0562e, PRG-1, mKIAA0455 |
257.5kb | Sense | Chr. 3 - 117319145 - 117360876 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

1700061I17Rik
Genomic Map