Assay Details
| Gene Symbol: Cdc42bpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033285.1 | NP_001028457.1 | 26 - 27 | 3423 | 90 | |
| GenBank mRNA | BC158017.1 | - | 26 - 27 | 3423 | 9007323 | 90 |
| BC158095.1 | - | 25 - 26 | 3384 | 9007382 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdc42bpa
Genomic Map