Assay Details
| Gene Symbol: Olfr414 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146761.2 | NP_666972.2 | 1 | 829 | 77 | |
| GenBank mRNA | BC106815.1 | - | 1 | 829 | 40043364 | 77 |
| BC106816.1 | - | 1 | 829 | 40043365 | 77 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P155258 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P124313 | NM_146761.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P124313 | BC106815.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P124313 | BC106816.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P124313 | NM_146761.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P124313 | BC106815.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P124313 | BC106816.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P124313 | NM_146761.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P124313 | BC106815.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P124313 | BC106816.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P124313 | NM_146761.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P124313 | BC106815.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P124313 | BC106816.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842226 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842244 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842254 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842249 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842215 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842262 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842243 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842269 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842240 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842264 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842266 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842223 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842268 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842218 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842230 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842232 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842228 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842242 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842234 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842257 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842263 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842222 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842221 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842259 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842224 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842260 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842231 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842251 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842237 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842235 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842220 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842256 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842246 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842267 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P155258 | - | - |
| Whole Mouse Genome G4122A | A_51_P124313 | NM_146761.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P124313 | BC106815.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P124313 | BC106816.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P124313 | NM_146761.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P124313 | BC106815.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P124313 | BC106816.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P107991 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102245 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P155258 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00776219 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00776215 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00776231 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00776229 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00776213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842254 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842226 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842246 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842208 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842240 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842264 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00492968 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00177016 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00452651 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842291 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842203 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842210 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842281 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00776222 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842268 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00177008 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842238 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842218 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842249 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842288 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842232 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842198 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842274 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842266 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842290 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00600506 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842262 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842222 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842199 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00776214 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842212 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00492970 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842214 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842221 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842223 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842224 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842276 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842273 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842244 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842271 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00178010 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00492966 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00776213 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842215 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842254 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842263 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842251 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842197 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842220 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00776220 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842231 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00776230 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842280 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842243 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00375755 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00177015 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00799773 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10351967 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10351966 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1740091 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001740091 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2600407 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2600407 | - | - |
| mouse-6 v1.1 (Illumina) | scl14314.1.1_14-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1740091 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl14314.1.1_14-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1740091 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2600407 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2600407 | - | - |

Olfr414
Genomic Map