Assay Details
| Gene Symbol: Frmd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190332.1 | NP_001177261.1 | 9 - 10 | 905 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Frmd7| Gene Symbol: Frmd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190332.1 | NP_001177261.1 | 9 - 10 | 905 | 90 | |
Set Membership: |
|