Assay Details
Gene Symbol: Tdrd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172605.3 | NP_766193.3 | 13 - 14 | 2471 | 64 | |
GenBank mRNA | AK033390.1 | - | 13 - 14 | 2416 | 64 | |
BB668608.1 | - | 2 - 3 | 165 | 64 | ||
BC005670.1 | - | 13 - 14 | 2372 | 3153274 | 64 | |
BC066144.1 | - | 13 - 14 | 2419 | 30435963 | 64 | |
BE983697.1 | - | 1 - 2 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
