Assay Details
Gene Symbol: Tdrd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253755.1 | NP_001240684.1 | 8 - 9 | 1086 | 78 | |
NM_172605.3 | NP_766193.3 | 8 - 9 | 1086 | 78 | ||
GenBank mRNA | AK032892.1 | - | 8 - 9 | 1031 | 78 | |
AK033390.1 | - | 8 - 9 | 1031 | 78 | ||
AK181483.1 | - | 2 - 3 | 282 | 78 | ||
BC005670.1 | - | 8 - 9 | 987 | 3153274 | 78 | |
BC066144.1 | - | 8 - 9 | 1034 | 30435963 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
