Assay Details
| Gene Symbol: Park2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016694.3 | NP_057903.1 | 2 - 3 | 269 | 93 | |
| GenBank mRNA | AB019558.1 | - | 2 - 3 | 240 | 93 | |
| AF250293.1 | - | 2 - 3 | 269 | 93 | ||
| AF250294.1 | - | 2 - 3 | 269 | 93 | ||
| AF250295.1 | - | 2 - 3 | 269 | 93 | ||
| BC113204.1 | - | 2 - 3 | 252 | 40047891 | 93 | |
| FJ555557.1 | - | 2 - 3 | 240 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Park2
Genomic Map