| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1400005838.mm.1 |
NM_080726.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400005838.mm.1 |
NM_080726.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400005838.mm.1 |
TC1400000748.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400005840.mm.1 |
NM_080726.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400005840.mm.1 |
NM_080726.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400005840.mm.1 |
TC1400000748.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400005835.mm.1 |
NM_080726.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400005835.mm.1 |
NM_080726.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400005835.mm.1 |
TC1400000748.mm.1 |
MTA TC |