Assay Details
| Gene Symbol: Rem2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080726.3 | NP_542764.2 | 2 - 3 | 622 | 64 | |
| GenBank mRNA | AK077317.1 | - | 2 - 3 | 503 | 64 | |
| AK090165.1 | - | 2 - 3 | 623 | 64 | ||
| BC018219.1 | - | 2 - 3 | 504 | 3155806 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rem2
Genomic Map