Assay Details
| Gene Symbol: Efr3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082483.1 | NP_001075952.1 | 15 - 16 | 1924 | 76 | |
| GenBank mRNA | BC075650.1 | - | 14 - 15 | 1681 | 76 | |
| BC147643.1 | - | 15 - 16 | 1924 | 9056633 | 76 | |
| BC157968.1 | - | 15 - 16 | 1924 | 9088136 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Efr3b
Genomic Map