Assay Details
Gene Symbol: Efr3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082483.1 | NP_001075952.1 | 16 - 17 | 2052 | 65 | |
GenBank mRNA | BC075650.1 | - | 15 - 16 | 1809 | 65 | |
BC147643.1 | - | 16 - 17 | 2052 | 9056633 | 65 | |
BC157968.1 | - | 16 - 17 | 2052 | 9088136 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
