Assay Details
| Gene Symbol: Pomc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008895.4 | NP_032921.1 | 1 - 2 | 101 | 60 | |
| GenBank mRNA | AK017492.1 | - | 1 - 2 | 97 | 60 | |
| AK133592.1 | - | 1 - 2 | 96 | 60 | ||
| AK133646.1 | - | 1 - 2 | 96 | 60 | ||
| AK133740.1 | - | 1 - 2 | 95 | 60 | ||
| AK133776.1 | - | 1 - 2 | 97 | 60 | ||
| AK133800.1 | - | 1 - 2 | 97 | 60 | ||
| BC061215.1 | - | 1 - 2 | 95 | 30253829 | 60 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK017492.1, AK133592.1, AK133646.1, AK133740.1, AK133776.1, AK133800.1, BC061215.1, NM_008895.4 transcripts.
Set Membership: |
|

Pomc
Genomic Map