Assay Details
| Gene Symbol: Spata17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028848.3 | NP_083124.1 | 3 - 4 | 446 | 97 | |
| NM_029067.1 | NP_083343.1 | 3 - 4 | 446 | 97 | ||
| GenBank mRNA | AY747687.1 | - | 3 - 4 | 421 | 97 | |
| BC089520.1 | - | 3 - 4 | 421 | 6774024 | 97 | |
More Information
Set Membership: |
|

Spata17
Genomic Map