Assay Details
| Gene Symbol: Khnyn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027143.2 | NP_081419.1 | 2 - 3 | 388 | 54 | |
| GenBank mRNA | AK033704.1 | - | 2 - 3 | 389 | 54 | |
| AK044902.1 | - | 2 - 3 | 361 | 54 | ||
| AK047955.1 | - | 2 - 3 | 357 | 54 | ||
| AK122247.1 | - | 2 - 3 | 340 | 54 | ||
| AK136301.1 | - | 2 - 3 | 415 | 54 | ||
| BC116260.1 | - | 2 - 3 | 375 | 40093805 | 54 | |
| BC116261.1 | - | 2 - 3 | 375 | 40093806 | 54 | |
More Information
Updated Mapping Information
Set Membership: |
|

Khnyn
Genomic Map