Assay Details
| Gene Symbol: Khnyn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027143.2 | NP_081419.1 | 3 - 4 | 1524 | 62 | |
| GenBank mRNA | AK032037.1 | - | 3 - 4 | 1496 | 62 | |
| AK033704.1 | - | 3 - 4 | 1525 | 62 | ||
| AK047054.1 | - | 2 - 3 | 2020 | 62 | ||
| AK047955.1 | - | 3 - 4 | 1493 | 62 | ||
| AK122247.1 | - | 3 - 4 | 1476 | 62 | ||
| AK136301.1 | - | 3 - 4 | 1551 | 62 | ||
| BC116260.1 | - | 3 - 4 | 1511 | 40093805 | 62 | |
| BC116261.1 | - | 3 - 4 | 1511 | 40093806 | 62 | |
More Information
Set Membership: |
|

Khnyn
Genomic Map