Assay Details
| Gene Symbol: Khnyn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027143.2 | NP_081419.1 | 6 - 7 | 1857 | 61 | |
| GenBank mRNA | AK032037.1 | - | 6 - 7 | 1829 | 61 | |
| AK033704.1 | - | 6 - 7 | 1858 | 61 | ||
| AK047054.1 | - | 6 - 7 | 2403 | 61 | ||
| AK047955.1 | - | 6 - 7 | 1826 | 61 | ||
| AK136301.1 | - | 6 - 7 | 1885 | 61 | ||
| BC116260.1 | - | 6 - 7 | 1844 | 40093805 | 61 | |
| BC116261.1 | - | 6 - 7 | 1844 | 40093806 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Khnyn
Genomic Map