Assay Details
| Gene Symbol: Rab30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029494.2 | NP_083770.2 | 1 - 2 | 133 | 131 | |
| GenBank mRNA | AK142206.1 | - | 1 - 2 | 140 | 131 | |
| BC017550.1 | - | 1 - 2 | 133 | 4503909 | 131 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK142206.1, BC017550.1, NM_029494.2 transcripts.
Set Membership: |
|

Rab30
Genomic Map