Assay Details
| Gene Symbol: Igsf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177591.4 | NP_808259.2 | 16 - 17 | 3309 | 63 | |
| NM_183336.2 | NP_899179.1 | 7 - 8 | 1563 | 63 | ||
| GenBank mRNA | AK030701.1 | - | 7 - 8 | 1565 | 63 | |
| AK076427.1 | - | 7 - 8 | 1556 | 63 | ||
| AY227771.1 | - | 16 - 17 | 3303 | 63 | ||
| AY227774.1 | - | 7 - 8 | 1558 | 63 | ||
| BC119264.1 | - | 7 - 8 | 1526 | 8734013 | 63 | |
| BC144762.1 | - | 7 - 8 | 1526 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Igsf1
Genomic Map