Assay Details
Gene Symbol: Abcb1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011075.2 | NP_035205.1 | 17 - 18 | 2348 | 125 | |
GenBank mRNA | AK030328.1 | - | 16 - 17 | 2291 | 125 | |
BC141363.1 | - | 17 - 18 | 2305 | 9055975 | 125 | |
BC150811.1 | - | 17 - 18 | 2305 | 9087722 | 125 | |
M14757.1 | - | 17 - 18 | 2308 | 125 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
