Assay Details
| Gene Symbol: Abcb1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011075.2 | NP_035205.1 | 4 - 5 | 428 | 71 | |
| GenBank mRNA | BC141363.1 | - | 4 - 5 | 385 | 9055975 | 71 |
| BC150811.1 | - | 4 - 5 | 385 | 9087722 | 71 | |
| BQ829721.1 | - | 3 - 4 | 317 | 71 | ||
| M14757.1 | - | 4 - 5 | 388 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Abcb1b
Genomic Map