Assay Details
| Gene Symbol: Igfl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003393.1 | NP_001003393.1 | 3 - 4 | 455 | 121 | |
| GenBank mRNA | AY672115.1 | - | 3 - 4 | 455 | 121 | |
| BC147097.1 | - | 3 - 4 | 455 | 9056138 | 121 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P120655 | NM_001003393.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P120655 | AY672115.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P120655 | BC147097.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P120655 | NM_001003393.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P120655 | AY672115.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P120655 | BC147097.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 340551 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 970613 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 939637 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 439073 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 48885 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P160000 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P120655 | NM_001003393.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P120655 | AY672115.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P120655 | BC147097.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P120655 | NM_001003393.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P120655 | AY672115.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P120655 | BC147097.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24987804 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24987789 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24987795 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24987769 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24987767 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24987781 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24987772 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24987793 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24987787 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24987786 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24987803 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24987802 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24987805 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24987776 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24987800 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24987778 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24987780 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24987788 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24987766 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24987779 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24987797 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24987764 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24987785 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24987783 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24987775 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24987791 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24987784 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24987794 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24987774 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24987790 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24987762 | - | - |
| Whole Mouse Genome G4122A | A_52_P120655 | NM_001003393.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P120655 | AY672115.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P120655 | BC147097.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P120655 | NM_001003393.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P120655 | AY672115.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P120655 | BC147097.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121628 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06527224 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05965459 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05965461 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05965463 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06527225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24987787 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24987770 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24987794 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24987772 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24987774 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24987768 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24987824 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24987787 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06295871 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24987800 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24987776 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06211514 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24987793 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24987750 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24987806 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24987780 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24987820 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24987788 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06528294 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24987810 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24987752 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05929057 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24987816 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24987817 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06211519 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24987821 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24987786 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24987798 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06365175 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05935946 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24987802 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24987807 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24987749 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06553699 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06095657 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24987769 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06636650 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24987760 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24987767 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24987778 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24987783 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24987762 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24987830 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24987785 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24987795 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06553709 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24987764 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06211518 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06527223 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06571274 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24987814 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24987827 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24987804 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06527224 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24987832 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06528298 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06578584 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10550483 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10550485 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10550484 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10550486 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10550482 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700001951.mm.1 | TC0700000264.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700001951.mm.1 | NM_001003393.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700001951.mm.1 | NM_001003393.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700001950.mm.1 | TC0700000264.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700001950.mm.1 | NM_001003393.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700001950.mm.1 | NM_001003393.1 | Refseq |
| Murine Genome U74v2 Set | 134738_at | - | - |
| Murine Genome U74v2 Set | 134738_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1242115 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2861641 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2861641 | - | - |

Igfl3
Genomic Map