Assay Details
| Gene Symbol: Wbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018765.3 | NP_061235.2 | 2 - 3 | 405 | 89 | |
| GenBank mRNA | AF071184.1 | - | 2 - 3 | 257 | 89 | |
| AK019160.1 | - | 2 - 3 | 139 | 89 | ||
| AK135215.1 | - | 2 - 3 | 405 | 89 | ||
| AK159522.1 | - | 2 - 3 | 657 | 89 | ||
| AK160559.1 | - | 2 - 3 | 142 | 89 | ||
| AK161496.1 | - | 2 - 3 | 99 | 89 | ||
| AK162009.1 | - | 2 - 3 | 147 | 89 | ||
| BC034851.1 | - | 2 - 3 | 100 | 1399984 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wbp4
Genomic Map