Assay Details
| Gene Symbol: Wbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018765.3 | NP_061235.2 | 7 - 8 | 898 | 91 | |
| GenBank mRNA | AF071184.1 | - | 7 - 8 | 750 | 91 | |
| AK135215.1 | - | 7 - 8 | 898 | 91 | ||
| AK159522.1 | - | 7 - 8 | 1150 | 91 | ||
| AK160559.1 | - | 7 - 8 | 635 | 91 | ||
| AK161496.1 | - | 7 - 8 | 592 | 91 | ||
| AK162009.1 | - | 7 - 8 | 640 | 91 | ||
| AK164984.1 | - | 1 - 2 | 2131 | 91 | ||
| BC034851.1 | - | 7 - 8 | 593 | 1399984 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wbp4
Genomic Map