Assay Details
| Gene Symbol: Wbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018765.3 | NP_061235.2 | 8 - 9 | 1088 | 73 | |
| GenBank mRNA | AK083381.1 | - | 1 - 2 | 143 | 73 | |
| AK135215.1 | - | 8 - 9 | 1088 | 73 | ||
| AK159522.1 | - | 8 - 9 | 1340 | 73 | ||
| AK160559.1 | - | 8 - 9 | 825 | 73 | ||
| AK161496.1 | - | 8 - 9 | 782 | 73 | ||
| AK162009.1 | - | 8 - 9 | 830 | 73 | ||
| AK164984.1 | - | 2 - 3 | 2321 | 73 | ||
| BC034851.1 | - | 8 - 9 | 783 | 1399984 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29751624 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29751626 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29751625 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29751622 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29751628 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29751623 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1998354 | AK135215.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1998354 | NM_018765.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 228243 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 534561 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 136793 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 107052 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 109011 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 522330 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 842252 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 39047 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 295194 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 530263 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 806868 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 334996 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 539529 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 235029 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 233963 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 103673 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 17097 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P178305 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P150192 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1998354 | AK135215.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1998354 | NM_018765.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P334604 | AK162009.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P334604 | AK164984.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P334604 | BC034851.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P334604 | AK159522.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P334604 | AK160559.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29751655 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29751644 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29751629 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29751659 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29751622 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29751652 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29751626 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29751627 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29751656 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29751642 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29751630 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29751639 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29751651 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29751634 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29751654 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29751625 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29751632 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29751623 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29751647 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29751628 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29751649 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29751637 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29751650 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29751646 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29751636 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P150192 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P178305 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08175565 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08175546 | - | - |
| Whole Mouse Genome G4122A | A_52_P572272 | AK083381.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P334604 | AK162009.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P334604 | AK164984.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P334604 | BC034851.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P334604 | AK159522.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P334604 | AK160559.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101895 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P178305 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08175546 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P150192 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11603335 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11603336 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11728232 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29751624 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29751628 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29751622 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29751626 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29751623 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29751625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751476 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751630 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11135966 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11603332 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751626 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751680 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751483 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751667 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751625 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751636 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751484 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11177222 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751656 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751646 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751642 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751679 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751663 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751655 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751654 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751662 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751639 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751671 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11818254 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11481319 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751681 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751657 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751637 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751652 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751651 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751628 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751624 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751644 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751486 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751634 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751682 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751669 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751659 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751650 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751487 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751670 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751666 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11481325 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751683 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751673 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10421851 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10421849 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10421843 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10421848 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10421846 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10421845 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10421842 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10421852 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10421841 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10421847 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10421850 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10421844 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1419375_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1419374_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10421840 | - | - |
| Mouse Genome 430 2.0 Array | 1419375_at | - | - |
| Mouse Genome 430 2.0 Array | 1419374_at | - | - |
| Mouse Genome 430 2.0 Array | 1419375_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1419374_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 22380_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 22380_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1400019148.mm.1 | NM_018765.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400019148.mm.1 | NM_018765.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400019148.mm.1 | TC1400002491.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400019152.mm.1 | NM_018765.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400019152.mm.1 | NM_018765.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400019152.mm.1 | TC1400002491.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400019149.mm.1 | NM_018765.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400019149.mm.1 | NM_018765.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400019149.mm.1 | TC1400002491.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1419374_PM_at | - | - |
| Mouse Expression Set 430 | 1419375_PM_at | - | - |
| Mouse Expression Set 430 | 1419374_at | - | - |
| Mouse Expression Set 430 | 1419375_at | - | - |
| Murine Genome U74v2 Set | 103836_at | - | - |
| Murine Genome U74v2 Set | 103836_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419374_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419374_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419375_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419375_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419374_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419375_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419375_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419374_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2230019 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2230019 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002230019 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1216130 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1216130 | - | - |
| mouse-6 v1.1 (Illumina) | scl45265.10.1_1-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2230019 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl45265.10.1_1-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2230019 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2462762 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2462762 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl45265.10.1_1-S | - | - |

Wbp4
Genomic Map