Assay Details
| Gene Symbol: Kbtbd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024135.2 | NP_001019306.2 | 1 | 4263 | 83 | |
| GenBank mRNA | AK003597.1 | - | 1 | 1516 | 83 | |
| BC096019.1 | - | 1 | 4263 | 6830769 | 83 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29751333 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29751335 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29751334 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P442852 | BC096019.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P442852 | NM_001024135.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P442852 | AK003597.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P442852 | BC096019.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P442852 | NM_001024135.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P442852 | AK003597.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 500285 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 177599 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P159599 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P442852 | BC096019.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P442852 | NM_001024135.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P442852 | AK003597.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P442852 | BC096019.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P442852 | NM_001024135.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P442852 | AK003597.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29751351 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29751335 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29751323 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29751353 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29751327 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29751330 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29751328 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29751320 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29751333 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29751344 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29751348 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29751319 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29751321 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29751345 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29751355 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29751325 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29751334 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P159599 | - | - |
| Whole Mouse Genome G4122A | A_52_P442852 | BC096019.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P442852 | NM_001024135.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P442852 | AK003597.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P442852 | BC096019.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P442852 | NM_001024135.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P442852 | AK003597.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P443952 | AK135854.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P443952 | BC096019.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P443952 | NM_001024135.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P159599 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11389756 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11232366 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11307811 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29751333 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29751334 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29751335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751320 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751345 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751326 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11307815 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751366 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751331 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751357 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11314324 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11314330 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751319 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11447814 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751320 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751374 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751371 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751328 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751344 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751355 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751349 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751372 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751377 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11249021 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751365 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751381 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11799781 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751333 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751361 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751369 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751383 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751358 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11307808 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11232365 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751378 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751375 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10416654 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452700_PM_s_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428192_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10416653 | - | - |
| Mouse Genome 430 2.0 Array | 1452700_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1428192_at | - | - |
| Mouse Genome 430 2.0 Array | 1428192_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1452700_PM_s_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 211255_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 211255_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1400008812.mm.1 | NM_001024135.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400008812.mm.1 | NM_001024135.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400008812.mm.1 | TC1400001103.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400008813.mm.1 | NM_001024135.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400008813.mm.1 | NM_001024135.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400008813.mm.1 | TC1400001103.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1452700_PM_s_at | - | - |
| Mouse Expression Set 430 | 1428192_at | - | - |
| Mouse Expression Set 430 | 1452700_s_at | - | - |
| Mouse Expression Set 430 | 1428192_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428192_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428192_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1452700_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1452700_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 630707 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2516565 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1240117 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3156633 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3077630 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3156633 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3077630 | - | - |

Kbtbd7
Genomic Map