Assay Details
| Gene Symbol: Bnc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007562.2 | NP_031588.2 | 3 - 4 | 438 | 73 | |
| GenBank mRNA | AK087567.1 | - | 3 - 4 | 432 | 73 | |
| U88064.1 | - | 3 - 4 | 385 | 73 | ||
More Information
Set Membership: |
|

Bnc1
Genomic Map