Assay Details
| Gene Symbol: Pde5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153422.2 | NP_700471.2 | 19 - 20 | 2461 | 122 | |
| GenBank mRNA | AF541937.1 | - | 19 - 20 | 2461 | 122 | |
| BC119137.1 | - | 19 - 20 | 2424 | 8733886 | 122 | |
| BC120486.1 | - | 19 - 20 | 2424 | 40129409 | 122 | |
More Information
Set Membership: |
|

Pde5a
Genomic Map