Assay Details
Gene Symbol: Pomt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145145.1 | NP_660127.1 | 1 - 2 | 63 | 90 | |
GenBank mRNA | AK053326.1 | - | 1 - 2 | 75 | 90 | |
AK053889.1 | - | 1 - 2 | 62 | 90 | ||
AK134770.1 | - | 1 - 2 | 107 | 90 | ||
AK153984.1 | - | 1 - 2 | 162 | 90 | ||
AK200298.1 | - | 1 - 2 | 41 | 90 | ||
AY007238.1 | - | 1 - 2 | 103 | 90 | ||
AY494857.1 | - | 1 - 2 | 123 | 90 | ||
BC027325.1 | - | 1 - 2 | 63 | 3981366 | 90 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK053889.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
