Assay Details
| Gene Symbol: Swi5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290552.1 | NP_001277481.1 | 4 - 5 | 310 | 128 | |
| NM_175190.5 | NP_780399.1 | 4 - 5 | 455 | 128 | ||
| NR_110965.1 | 3 - 4 | 330 | 128 | |||
| GenBank mRNA | AK013517.1 | - | 4 - 5 | 415 | 128 | |
| AK018775.1 | - | 3 - 4 | 268 | 128 | ||
| AK041567.1 | - | 4 - 5 | 397 | 128 | ||
| AK088274.1 | - | 4 - 5 | 416 | 128 | ||
| AK166608.1 | - | 4 - 5 | 399 | 128 | ||
| AK216650.1 | - | 2 - 3 | 224 | 128 | ||
| BC021748.1 | - | 4 - 5 | 405 | 3586619 | 128 | |
| BY683904.1 | - | 1 - 2 | 77 | 128 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21114432 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21114426 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21114431 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21114429 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21114428 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21114427 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2040046 | AK166608.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2040046 | BC021748.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2040046 | BY683904.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2040046 | NM_175190.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2040046 | AK013517.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2040048 | AK166608.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2040048 | AK216650.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2040048 | BY683904.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2040048 | NM_175190.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2040048 | AK013517.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 395111 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 928658 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 466635 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 614640 | AK089251.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 845280 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 14771 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P119937 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2040046 | AK166608.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2040046 | BC021748.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2040046 | BY683904.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2040046 | NM_175190.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2040046 | AK013517.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2040048 | AK166608.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2040048 | AK216650.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2040048 | BY683904.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2040048 | NM_175190.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2040048 | AK013517.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21114422 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21114442 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21114420 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21114438 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21114458 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21114429 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21114451 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21114427 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21114419 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21114440 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21114450 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21114428 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21114454 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21114424 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21114456 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21114455 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21114433 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21114435 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21114437 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21114459 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21114426 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21114436 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21114431 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21114445 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21114448 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21114453 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00337729 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P119937 | - | - |
| Whole Mouse Genome G4122A | A_52_P599181 | AK166608.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P599181 | AK216650.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P599181 | BY683904.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P599181 | NM_175190.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P599181 | AK013517.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P444401 | AK039840.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P599175 | AK166608.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P599175 | AK216650.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P599175 | BC021748.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P599175 | BY683904.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P599175 | NM_175190.4 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135915 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P119937 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01596938 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01596949 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01596940 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01596956 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01596943 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21114427 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21114426 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21114431 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21114432 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21114429 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21114428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21114429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21114399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21114382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21114395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21114394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21114355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21114426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21114371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21114361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21114390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21114427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21114378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21114381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21114375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21114373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21114397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21114351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21114409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21114422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21114385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21114431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21114363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21114419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21114401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21114406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21114412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21114400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21114387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21114415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21114393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21114404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21114402 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21114368 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21114404 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21114361 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21114413 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02224977 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21114426 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21114409 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01596943 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21114424 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21114417 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21114410 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01596947 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21114364 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21114407 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21114415 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21114412 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21114420 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21114401 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01913558 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21114421 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02252383 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21114422 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21114402 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01312871 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21114419 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21114363 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10481586 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10481587 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10481588 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10481589 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10481591 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1433480_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10481585 | - | - |
| Mouse Genome 430 2.0 Array | 1433480_at | - | - |
| Mouse Genome 430 2.0 Array | 1433480_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72931_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72931_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200028001.mm.1 | NM_175190.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200028001.mm.1 | TC0200003299.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200028000.mm.1 | NM_175190.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200028000.mm.1 | TC0200003299.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1433480_at | - | - |
| Mouse Expression Set 430 | 1433480_PM_at | - | - |
| Murine Genome U74v2 Set | 96658_PM_at | - | - |
| Murine Genome U74v2 Set | 96658_at | - | - |
| Mouse Genome 430A 2.0 Array | 1433480_at | - | - |
| Mouse Genome 430A 2.0 Array | 1433480_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1433480_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1433480_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 510333 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 510333 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000510333 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2733835 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2733835 | - | - |
| mouse-6 v1.1 (Illumina) | scl19447.7.1_15-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 510333 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl19447.7.1_15-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 510333 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2733835 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2733835 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl19447.7.1_15-S | - | - |

Swi5
Genomic Map