Assay Details
| Gene Symbol: Rtf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030112.2 | NP_084388.2 | 3 - 4 | 470 | 105 | |
| GenBank mRNA | AK019305.1 | - | 2 - 3 | 215 | 105 | |
| BC022717.1 | - | 4 - 5 | 479 | 105 | ||
| BC038032.1 | - | 3 - 4 | 443 | 105 | ||
| BC048535.1 | - | 3 - 4 | 454 | 105 | ||
| BC117848.1 | - | 3 - 4 | 322 | 40093717 | 105 | |
| BC117849.1 | - | 3 - 4 | 322 | 40093724 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rtf1
Genomic Map