Assay Details
Gene Symbol: Rtf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030112.2 | NP_084388.2 | 8 - 9 | 1219 | 69 | |
GenBank mRNA | AK135633.1 | - | 2 - 3 | 287 | 69 | |
BC117848.1 | - | 8 - 9 | 1071 | 40093717 | 69 | |
BC117849.1 | - | 8 - 9 | 1071 | 40093724 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
